Assay Details
| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013576.4 | XP_004013625.1 | 1 - 2 | 1906 | 95 | |
| XM_012186971.3 | XP_012042361.1 | 2 - 3 | 763 | 95 | ||
More Information
Set Membership: |
|

YOD1
Genomic Map