Assay Details
| Gene Symbol: LOC101116026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012111867.3 | XP_011967257.2 | 2 - 3 | 1775 | 57 | |
| XM_012111874.3 | XP_011967264.2 | 2 - 3 | 1776 | 57 | ||
| XM_012111887.3 | XP_011967277.1 | 2 - 3 | 496 | 57 | ||
More Information
Set Membership: |
|

LOC101116026
Genomic Map