Assay Details
| Gene Symbol: MOGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012147009.2 | XP_012002399.2 | 4 - 5 | 673 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MOGAT1| Gene Symbol: MOGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012147009.2 | XP_012002399.2 | 4 - 5 | 673 | 122 | |
Set Membership: |
|