Assay Details
| Gene Symbol: CRYBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965472.1 | XP_027821273.1 | 6 - 7 | 3920 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRYBG2| Gene Symbol: CRYBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965472.1 | XP_027821273.1 | 6 - 7 | 3920 | 98 | |