Assay Details
| Gene Symbol: LRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978187.1 | XP_027833988.1 | 5 - 6 | 1726 | 68 | |
| XM_027978188.1 | XP_027833989.1 | 5 - 6 | 1690 | 68 | ||
| XM_027978189.1 | XP_027833990.1 | 5 - 6 | 1629 | 68 | ||
| XM_027978190.1 | XP_027833991.1 | 5 - 6 | 1593 | 68 | ||
| XM_027978191.1 | XP_027833992.1 | 5 - 6 | 1732 | 68 | ||
More Information
Set Membership: |
|

LRP3
Genomic Map