Assay Details
| Gene Symbol: TMEM72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962272.1 | XP_027818073.1 | 2 - 3 | 1882 | 108 | |
| XM_027962273.1 | XP_027818074.1 | 3 - 4 | 405 | 108 | ||
More Information
Set Membership: |
|

TMEM72
Genomic Map