Assay Details
| Gene Symbol: LOC101114349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012176323.3 | XP_012031713.2 | 1 - 2 | 1080 | 95 | |
| XM_012176324.3 | XP_012031714.2 | 1 - 2 | 1080 | 95 | ||
| XM_027968443.1 | XP_027824244.1 | 1 - 2 | 1080 | 95 | ||
| XM_027968444.1 | XP_027824245.1 | 1 - 2 | 1080 | 95 | ||

LOC101114349
Genomic Map