Assay Details
Gene Symbol: LOC101114349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012176323.3 | XP_012031713.2 | 4 - 5 | 1502 | 103 | |
XM_012176324.3 | XP_012031714.2 | 4 - 5 | 1502 | 103 | ||
XM_027968443.1 | XP_027824244.1 | 4 - 5 | 1502 | 103 | ||
XM_027968444.1 | XP_027824245.1 | 4 - 5 | 1502 | 103 | ||
XM_027968445.1 | XP_027824246.1 | 3 - 4 | 1346 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
