Assay Details
| Gene Symbol: LOC101120574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969284.1 | XP_027825085.1 | 4 - 5 | 538 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120574| Gene Symbol: LOC101120574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969284.1 | XP_027825085.1 | 4 - 5 | 538 | 64 | |
Set Membership: |
|