Assay Details
| Gene Symbol: DUSP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098810.2 | XP_014954296.1 | 1 - 2 | 200 | 96 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUSP3| Gene Symbol: DUSP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098810.2 | XP_014954296.1 | 1 - 2 | 200 | 96 | |