Assay Details
| Gene Symbol: LOC101118398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092330.2 | XP_014947816.1 | 8 - 9 | 1368 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101118398| Gene Symbol: LOC101118398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092330.2 | XP_014947816.1 | 8 - 9 | 1368 | 70 | |
Set Membership: |
|