Assay Details
| Gene Symbol: LOC101122595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177290.2 | XP_012032680.2 | 1 - 2 | 136 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122595| Gene Symbol: LOC101122595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177290.2 | XP_012032680.2 | 1 - 2 | 136 | 97 | |
Set Membership: |
|