Assay Details
Gene Symbol: LOC101122595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177290.2 | XP_012032680.2 | 3 - 4 | 360 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101122595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177290.2 | XP_012032680.2 | 3 - 4 | 360 | 143 |
Set Membership: |
![]() ![]() ![]() |