Assay Details
| Gene Symbol: SSC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978773.1 | XP_027834574.1 | 12 - 13 | 2905 | 62 | |
| XM_027978774.1 | XP_027834575.1 | 12 - 13 | 2902 | 62 | ||
More Information
Set Membership: |
|

SSC5D
Genomic Map