Assay Details
| Gene Symbol: LOC101116255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979931.1 | XP_027835732.1 | 8 - 9 | 935 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101116255| Gene Symbol: LOC101116255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979931.1 | XP_027835732.1 | 8 - 9 | 935 | 63 | |
Set Membership: |
|