Assay Details
| Gene Symbol: LOC101122710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975403.1 | XP_027831204.1 | 1 - 2 | 34 | 64 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122710| Gene Symbol: LOC101122710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975403.1 | XP_027831204.1 | 1 - 2 | 34 | 64 | |