Assay Details
| Gene Symbol: LOC101119839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976439.1 | XP_027832240.1 | 2 - 3 | 429 | 61 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101119839| Gene Symbol: LOC101119839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976439.1 | XP_027832240.1 | 2 - 3 | 429 | 61 | |
Set Membership: | 
		   
					
						
							 |