Assay Details
Gene Symbol: LOC101121054 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003092.4 | XP_004003141.1 | 18 - 19 | 2498 | 94 | |
XM_027957697.1 | XP_027813498.1 | 17 - 18 | 2094 | 94 | ||
XM_027957708.1 | XP_027813509.1 | 16 - 17 | 2007 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
