Assay Details
| Gene Symbol: ZDHHC21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004383.4 | XP_004004432.1 | 6 - 7 | 831 | 85 | |
| XM_012125908.3 | XP_011981298.1 | 6 - 7 | 804 | 85 | ||
| XM_012125926.3 | XP_011981316.1 | 5 - 6 | 655 | 85 | ||
| XM_012125943.3 | XP_011981333.1 | 6 - 7 | 841 | 85 | ||
| XM_027964561.1 | XP_027820362.1 | 5 - 6 | 538 | 85 | ||
More Information
Set Membership: |
|

ZDHHC21
Genomic Map