Assay Details
| Gene Symbol: RETREG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004940.4 | XP_004004989.2 | 8 - 9 | 1104 | 76 | |
| XM_027965206.1 | XP_027821007.1 | 7 - 8 | 580 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RETREG2| Gene Symbol: RETREG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004940.4 | XP_004004989.2 | 8 - 9 | 1104 | 76 | |
| XM_027965206.1 | XP_027821007.1 | 7 - 8 | 580 | 76 | ||