Assay Details
| Gene Symbol: ADIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007604.3 | XP_004007653.2 | 7 - 8 | 1239 | 73 | |
| GenBank mRNA | KF921623.1 | - | 7 - 8 | 1229 | 73 | |
More Information
Set Membership: |
|

ADIPOR2
Genomic Map