Assay Details
| Gene Symbol: LOC114116347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972870.1 | XP_027828671.1 | 5 - 6 | 563 | 89 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114116347| Gene Symbol: LOC114116347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972870.1 | XP_027828671.1 | 5 - 6 | 563 | 89 | |