Assay Details
| Gene Symbol: LOC101118120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975898.1 | XP_027831699.1 | 14 - 15 | 1965 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101118120| Gene Symbol: LOC101118120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975898.1 | XP_027831699.1 | 14 - 15 | 1965 | 75 | |