Assay Details
| Gene Symbol: PCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969718.1 | XP_027825519.1 | 3 - 4 | 533 | 62 | |
| XM_027969719.1 | XP_027825520.1 | 3 - 4 | 528 | 62 | ||
| XM_027969720.1 | XP_027825521.1 | 3 - 4 | 512 | 62 | ||
More Information
Set Membership: |
|

PCP2
Genomic Map