Assay Details
| Gene Symbol: LOC114112016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963702.1 | XP_027819503.1 | 1 - 2 | 135 | 115 | |
| Gene Symbol: SWT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187729.3 | XP_012043119.1 | 12 - 13 | 2030 | 115 | |
| XM_012187733.3 | XP_012043123.1 | 12 - 13 | 2030 | 115 | ||
More Information
Set Membership: |
|

LOC114112016
Genomic Map