Assay Details
| Gene Symbol: NEMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093322.2 | XP_014948808.2 | 7 - 8 | 1042 | 81 | |
| XM_027964705.1 | XP_027820506.1 | 7 - 8 | 3773 | 81 | ||
| XM_027964706.1 | XP_027820507.1 | 8 - 9 | 1056 | 81 | ||

NEMP2
Genomic Map