Assay Details
| Gene Symbol: LOC101114669 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963782.1 | XP_027819583.1 | 6 - 7 | 816 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101114669| Gene Symbol: LOC101114669 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963782.1 | XP_027819583.1 | 6 - 7 | 816 | 60 | |
Set Membership: |
|