Assay Details
| Gene Symbol: UBAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004135.4 | XP_004004184.1 | 5 - 6 | 1495 | 70 | |
| XM_012118171.3 | XP_011973561.1 | 5 - 6 | 1783 | 70 | ||
| XM_012118181.3 | XP_011973571.1 | 4 - 5 | 1658 | 70 | ||
| XM_015093072.2 | XP_014948558.1 | 4 - 5 | 1363 | 70 | ||
More Information
Set Membership: |
|

UBAP1
Genomic Map