Assay Details
| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013048.4 | XP_004013097.3 | 11 - 12 | 1257 | 83 | |
| XM_027974313.1 | XP_027830114.1 | 11 - 12 | 1257 | 83 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013048.4 | XP_004013097.3 | 11 - 12 | 1257 | 83 | |
| XM_027974313.1 | XP_027830114.1 | 11 - 12 | 1257 | 83 | ||