Assay Details
| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013048.4 | XP_004013097.3 | 8 - 9 | 905 | 97 | |
| XM_027974312.1 | XP_027830113.1 | 8 - 9 | 905 | 97 | ||
| XM_027974313.1 | XP_027830114.1 | 8 - 9 | 905 | 97 | ||

HEXD
Genomic Map