Assay Details
| Gene Symbol: TXNL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015109.4 | XP_004015158.1 | 3 - 4 | 580 | 143 | |
| XM_004015110.4 | XP_004015159.1 | 4 - 5 | 711 | 143 | ||
| XM_027978136.1 | XP_027833937.1 | 4 - 5 | 909 | 143 | ||
More Information
Set Membership: |
|

TXNL4B
Genomic Map