Assay Details
| Gene Symbol: LOC101106373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018986.4 | XP_004019035.1 | 6 - 7 | 1188 | 119 | |
| XM_027958869.1 | XP_027814670.1 | 7 - 8 | 1274 | 119 | ||
| XM_027958870.1 | XP_027814671.1 | 6 - 7 | 1218 | 119 | ||
| XM_027958871.1 | XP_027814672.1 | 6 - 7 | 1149 | 119 | ||
More Information
Set Membership: |
|

LOC101106373
Genomic Map