Assay Details
| Gene Symbol: LOC101104287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103582.2 | XP_014959068.1 | 6 - 7 | 730 | 84 | |
| XM_027960758.1 | XP_027816559.1 | 6 - 7 | 832 | 84 | ||
More Information
Set Membership: |
|

LOC101104287
Genomic Map