Assay Details
| Gene Symbol: SEPTIN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961725.1 | XP_027817526.1 | 2 - 3 | 290 | 84 | |
| XM_027961726.1 | XP_027817527.1 | 2 - 3 | 290 | 84 | ||
More Information
Set Membership: |
|

SEPTIN12
Genomic Map