Assay Details
| Gene Symbol: TBCEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095622.3 | XP_011951012.1 | 6 - 7 | 1361 | 66 | |
| XM_027979333.1 | XP_027835134.1 | 6 - 7 | 939 | 66 | ||
More Information
Set Membership: |
|

TBCEL
Genomic Map