Assay Details
| Gene Symbol: SCNN1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104264.3 | XP_011959654.1 | 10 - 11 | 1543 | 64 | |
| GenBank mRNA | AF250862.1 | - | 7 - 8 | 64 | ||
More Information
Set Membership: |
|

SCNN1G
Genomic Map