Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974155.1 | XP_027829956.1 | 3 - 4 | 772 | 52 | |
| XM_027974156.1 | XP_027829957.1 | 2 - 3 | 900 | 52 | ||
| XM_027974157.1 | XP_027829958.1 | 2 - 3 | 771 | 52 | ||
| XM_027974158.1 | XP_027829959.1 | 2 - 3 | 679 | 52 | ||
More Information
Set Membership: |
|

SEPTIN9
Genomic Map