Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974155.1 | XP_027829956.1 | 6 - 7 | 1175 | 97 | |
| XM_027974156.1 | XP_027829957.1 | 5 - 6 | 1303 | 97 | ||
| XM_027974157.1 | XP_027829958.1 | 5 - 6 | 1174 | 97 | ||
| XM_027974158.1 | XP_027829959.1 | 5 - 6 | 1082 | 97 | ||
| XM_027974159.1 | XP_027829960.1 | 4 - 5 | 573 | 97 | ||
| XM_027974160.1 | XP_027829961.1 | 4 - 5 | 434 | 97 | ||
| XM_027974161.1 | XP_027829962.1 | 4 - 5 | 518 | 97 | ||
| XM_027974162.1 | XP_027829963.1 | 4 - 5 | 610 | 97 | ||
| XM_027974163.1 | XP_027829964.1 | 4 - 5 | 887 | 97 | ||
| XM_027974164.1 | XP_027829965.1 | 4 - 5 | 457 | 97 | ||
More Information
Set Membership: |
|

SEPTIN9
Genomic Map