Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974155.1 | XP_027829956.1 | 10 - 11 | 1623 | 84 | |
| XM_027974156.1 | XP_027829957.1 | 9 - 10 | 1751 | 84 | ||
| XM_027974157.1 | XP_027829958.1 | 9 - 10 | 1622 | 84 | ||
| XM_027974158.1 | XP_027829959.1 | 9 - 10 | 1530 | 84 | ||
| XM_027974159.1 | XP_027829960.1 | 8 - 9 | 1021 | 84 | ||
| XM_027974160.1 | XP_027829961.1 | 8 - 9 | 882 | 84 | ||
| XM_027974161.1 | XP_027829962.1 | 8 - 9 | 966 | 84 | ||
| XM_027974162.1 | XP_027829963.1 | 8 - 9 | 1058 | 84 | ||
| XM_027974163.1 | XP_027829964.1 | 8 - 9 | 1335 | 84 | ||
| XM_027974164.1 | XP_027829965.1 | 8 - 9 | 905 | 84 | ||
More Information
Set Membership: |
|

SEPTIN9
Genomic Map