Assay Details
| Gene Symbol: MAPRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099832.2 | XP_014955318.1 | 6 - 7 | 847 | 73 | |
| XM_027977074.1 | XP_027832875.1 | 6 - 7 | 834 | 73 | ||
| XM_027977075.1 | XP_027832876.1 | 6 - 7 | 834 | 73 | ||
More Information
Set Membership: |
|

MAPRE1
Genomic Map