Assay Details
| Gene Symbol: LOC443459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180067.3 | XP_012035457.2 | 1 - 2 | 139 | 80 | |
| XM_012180068.3 | XP_012035458.2 | 1 - 2 | 140 | 80 | ||
More Information
Set Membership: |
|

LOC443459
Genomic Map