Assay Details
| Gene Symbol: SERPINB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020581.3 | XP_004020630.3 | 5 - 6 | 563 | 77 | |
| XM_027960876.1 | XP_027816677.1 | 5 - 6 | 563 | 77 | ||
More Information
Set Membership: |
|

SERPINB10
Genomic Map