Assay Details
Gene Symbol: LOC101110623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012098697.2 | XP_011954087.2 | 5 - 6 | 582 | 92 | |
XM_012098699.3 | XP_011954089.1 | 5 - 6 | 575 | 92 | ||
XM_027957191.1 | XP_027812992.1 | 5 - 6 | 582 | 92 | ||
XM_027957192.1 | XP_027812993.1 | 5 - 6 | 575 | 92 | ||
XM_027957193.1 | XP_027812994.1 | 5 - 6 | 582 | 92 |
