Assay Details
Gene Symbol: LOC101110623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012098697.2 | XP_011954087.2 | 9 - 10 | 942 | 102 | |
XM_012098699.3 | XP_011954089.1 | 9 - 10 | 935 | 102 | ||
XM_027957191.1 | XP_027812992.1 | 10 - 11 | 1110 | 102 | ||
XM_027957192.1 | XP_027812993.1 | 10 - 11 | 1103 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
