Assay Details
| Gene Symbol: FRMD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012181347.3 | XP_012036737.1 | 9 - 10 | 1113 | 67 | |
| XM_027971739.1 | XP_027827540.1 | 10 - 11 | 1141 | 67 | ||
| XM_027971740.1 | XP_027827541.1 | 9 - 10 | 1089 | 67 | ||
| XM_027971741.1 | XP_027827542.1 | 8 - 9 | 866 | 67 | ||
More Information
Set Membership: |
|

FRMD6
Genomic Map