Assay Details
| Gene Symbol: LOC101110605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012186735.3 | XP_012042125.3 | 3 - 4 | 387 | 85 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101110605| Gene Symbol: LOC101110605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012186735.3 | XP_012042125.3 | 3 - 4 | 387 | 85 | |