Assay Details
| Gene Symbol: LOC114116366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972982.1 | XP_027828783.1 | 6 - 7 | 461 | 69 | |
| Gene Symbol: NSMAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972974.1 | XP_027828775.1 | 11 - 12 | 932 | 69 | |
More Information
Set Membership: |
|

LOC114116366
Genomic Map