Assay Details
| Gene Symbol: LOC114116419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973280.1 | XP_027829081.1 | 3 - 4 | 364 | 85 | |
| Gene Symbol: NSMAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972974.1 | XP_027828775.1 | 15 - 16 | 1391 | 85 | |

LOC114116419
Genomic Map