Assay Details
| Gene Symbol: LOC114116366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972982.1 | XP_027828783.1 | 4 - 5 | 259 | 74 | |
| Gene Symbol: NSMAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972974.1 | XP_027828775.1 | 9 - 10 | 730 | 74 | |

LOC114116366
Genomic Map