Assay Details
| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977694.1 | XP_027833495.1 | 6 - 7 | 674 | 89 | |
| XM_027977695.1 | XP_027833496.1 | 6 - 7 | 690 | 89 | ||
| XM_027977696.1 | XP_027833497.1 | 6 - 7 | 703 | 89 | ||
| XM_027977697.1 | XP_027833498.1 | 6 - 7 | 675 | 89 | ||
More Information
Set Membership: |
|

CIBAR2
Genomic Map